Learn More
Novus Biologicals™ Bone marrow stromal cell antigen 1/CD157 Antibody Pair [HRP]
Shop All Novus Biologicals Products

Description
Specifications
Specifications
| Assay Range | 23.44 pg/mL |
| Assay Sensitivity | 23.44-1500 pg/mL |
| Assay | Sandwich ELISA |
| Conjugate | HRP |
| Product Type | Antibody Pairs |
| Specificity | This solid phase sandwich ELISA utilizes an antibody specific for Human Bone marrow stromal cell antigen 1/CD157. |
| Target Species | Human |
| For Use With (Application) | Sandwich ELISA |
| Gene Alias | ADP-ribosyl cyclase 2, Bone marrow stromal antigen 1, bone marrow stromal cell antigen 1, BST-1, cADPr hydrolase 2, CD157, CD157 antigen, Cyclic ADP-ribose hydrolase 2, EC 3.2.2.5, NAD(+) nucleosidase |
| Gene ID (Entrez) | 683 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.