Learn More
Creative Biomart Bone Morphogenetic Protein-13 (BMP-13) Protein
Supplier:  Creative Biomart THP0254
Encompass_Preferred
This 27.5 kDa protein is obtained by proteolytic processing of a biologically inactive precursor protein of 97.7 kDa. Recombinant Human BMP-13/CDMP-2 is a 27.0 kDa homodimeric disulfide-linked protein consisting of two 120 amino acid polypeptide chains.
Catalog No. 50-208-4305
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.