Learn More
Sigma Aldrich Fine Chemicals Biosciences Bone Morphogenetic Protein
Supplier:  Sigma Aldrich Fine Chemicals Biosciences H479110UG
Encompass
Bone Morphogenetic Protein 2 is a member of the TGf-beta superfamily of cytokines that affect bone and cartilage formation. It is important for skeletal development during embryogenesis. BMP-2 induces chondrocyte formation osteoblast differentiation and is involved in embryo dorsal-ventral patterning and organogenesis.
Catalog No. 50-290-4649
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.