Learn More
Amsbio LLC Multiple myeloma (MM) and extramedullary plasmacytoma (EMP) with bone marrow tissue array
Supplier:  Amsbio LLC BM483D
Encompass
Bone tumor and bone marrow tissue array containing plasma cell myeloma (MM) primitive neuroectodermal tumor(PNET) Ewing's sarcoma and diffuse large B-cell lymphoma24 cases/48 cores (core size 1.5mm) replacing BM483c
Catalog No. 50-256-4549