Description
- Tungsten carbide is 5 times harder than stainless steel.
- Scissors with tungsten carbide inserts along their blades last much longer than stainless steel scissors.
Specifications
Specifications
| For Use With (Application) | Neuroscience |
| Length (English) | 3.5 in. |
| Material | Tungsten |
| Model | Straight, Strong cut |