Learn More
Description
- Designed to support increased upstream viral vector productivity in HEK293 cell lines
- Used together with boost expression supplement a as a defined supplement pair to optimize expression performance
- Alkaline, concentrated solution of amino acids
- Chemically defined, animal component-free formulation
- Ready-to-use liquid
- Suitable for integration into existing upstream processes without disruptive changes
- Consistent performance from development through production scale
- Manufactured at ISO 13485 certified manufacturing sites
Specifications
Specifications
| Classification | Chemically-defined, Animal Origin-free |
| Form | Liquid |
| Product Type | Cell Culture Supplement |
| Sterility | Non-sterile |
| With Additives | None |
| Without Additives | None |
| Quantity | 1000 mL |
| Regulatory Status | Non-regulated |
For further manufacturing or research use only. Not for diagnostic or therapeutic use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.