Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Borane - 2-Methylpyridine Complex 85.0+%, TCI America™
SDP

Catalog No. B30185G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30185G 5 g
B301825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B30185G Supplier TCI America Supplier No. B30185G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 3999-38-0 | C6H10BN | 106.96 g/mol

Chemical Identifiers

CAS 3999-38-0
Molecular Formula C6H10BN
Molecular Weight (g/mol) 106.96
MDL Number MFCD07784361
InChI Key ZMASBEFBIXMNCP-UHFFFAOYSA-N
Synonym Borane - 2-Picoline Complex, 2-Picoline Borane, 2-Methylpyridine Borane
IUPAC Name 2-methylpyridine borane
SMILES B.CC1=CC=CC=N1

Specifications

Melting Point 48°C
Color White-Yellow
Quantity 5 g
UN Number 2813
Formula Weight 106.96
Percent Purity ≥85.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Borane - 2-Methylpyridine Complex
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.