Learn More
Borane-dimethylamine complex, 97%
Description
Borane-dimethylamine complex is used in a nickel-catalyzed reduction of electron-deficient aryl tosylates. It also serves as a reducing agent in electrolytic deposition of metals, alloys, semiconductors and insulators.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 36°C |
Color | White |
Density | 0.69 g/mL |
Boiling Point | 59°C to 65°C (1 to 2 mmHg) |
Quantity | 100 g |
UN Number | UN2925 |
Sensitivity | Air and moisture sensitive |
Solubility Information | Soluble in methanol,water,ethanol,2-propanol and terahydrofuran. |
Formula Weight | 58.92 |
Percent Purity | 97% |
Show More |
Safety and Handling
Hazard Category | H228-H301+H311-H315-H319-H332-H335 |
Hazard Statement | GHS H Statement H301-H310-H330-H314-H318-H228 Toxic if swallowed. Fatal in contact with skin. Fatal if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage. Flammable solid. |
Precautionary Statement | P210-P240-P241-P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P370+P378q-P501c |
DOTInformation | Hazard Class: 4.1; Packaging Group: II |
EINECSNumber | 200-823-7 |
RTECSNumber | IP9450000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.