Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Borane-methyl sulfide complex, 1M solution in methylene chloride, AcroSeal™, Thermo Scientific Chemicals

Catalog No. AC185618000
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC185618000 800 mL Glass Bottle
AC185611000 100 mL AcroSeal™ Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC185618000 Supplier Thermo Scientific Chemicals Supplier No. 185618000

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13292-87-0 , 75-09-2
Molecular Formula C2H9BS
Molecular Weight (g/mol) 75.96
MDL Number MFCD00013189
InChI Key RMHDLBZYPISZOI-UHFFFAOYSA-N
Synonym dimethyl sulfide borane, borane-methyl sulfide complex, borane dimethyl sulfide complex, borane-dimethyl sulfide, dimethyl sulfideborane, dimethyl sulfide trihydroboron, borane-dimethyl sulfide complex, borane methylsulfide, borane dimethylsulfide, borane dms
PubChem CID 9833925
SMILES B.CSC

Specifications

Color Colorless
Linear Formula (CH3)2S·BH3
Concentration 0.95 to 1.05M
Merck Index 15, 1336
IUPAC Name (methylsulfanyl)methane borane
Formula Weight 75.95
Packaging Glass Bottle
Quantity 800 mL
Physical Form Liquid
Chemical Name or Material Borane-methyl sulfide complex, 1M Solutionin Methylene Chloride
Hazard Category Danger
Hazard Statement GHS H Statement
In contact with water releases flammable gases which may ignite spontaneously. Harmful if swallowed. Causes skin irritation. Causes serious eye damage. May cause drowsiness or dizziness. Suspected of causing cancer if inhaled. Reacts violently with water.
Precautionary Statement GHS P Statement
Handle under inert gas. Protect from moisture. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
EINECSNumber 236-313-6
RTECSNumber PV5080000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.