Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Borane-morpholine complex, 97%

Catalog No. AAL1511006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1511006 5 g
AAL1511014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1511006 Supplier Thermo Scientific Chemicals Supplier No. L1511006
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 4856-95-5 | C4H12BNO | 100.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4856-95-5
Molecular Formula C4H12BNO
Molecular Weight (g/mol) 100.96
MDL Number MFCD00066736
InChI Key IFVAHVOGOBRFSP-UHFFFAOYSA-N
Synonym morpholineborane, morpholine borane, borane morpholine complex, borane-morpholine complex, c4h12bno, morpholin-boran, borane morpholine, borane, compound with morpholine, boron; morpholine
PubChem CID 6327109
IUPAC Name boron;morpholine
SMILES B.C1COCCN1

Specifications

Melting Point 93°C to 95°C
Odor Amine-like
Quantity 5 g
UN Number UN2925
Formula Weight 100.96
Percent Purity 97%
Chemical Name or Material Borane-morpholine complex
Hazard Category H228-H302-H312-H314-H335-H500
Hazard Statement GHS H Statement
H228-H301-H314-H318
Flammable solid.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P240-P241-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, CORROSIVE, ORGANIC, N.O.S.
EINECSNumber 225-450-7
RTECSNumber QD8530000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.