Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Borane-trimethylamine complex, 97%

Catalog No. AC402740250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC402740250 25 g Glass bottle
AC402741000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC402740250 Supplier Thermo Scientific Chemicals Supplier No. 402740250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
This item is not returnable. View return policy

CAS: 75-22-9 | C3H12BN | 72.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-22-9
Molecular Formula C3H12BN
Molecular Weight (g/mol) 72.95
MDL Number MFCD00012420
InChI Key WVMHLYQJPRXKLC-UHFFFAOYSA-N
Synonym Trimethylamine borane
IUPAC Name trimethylamine borane
SMILES B.CN(C)C

Specifications

Melting Point 92.0°C to 96.0°C
Color White
Boiling Point 172.0°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (CH3)3N·BH3
Quantity 25 g
Fieser 01,1229; 05,708; 12,65
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in ethanols,ethyl ether
Formula Weight 72.95
Percent Purity 97%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material Borane-trimethylamine complex, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Flammable solid.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-850-4
RTECSNumber PA0525000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.