Learn More
Boston Bioproducts Inc Borate Buffer (0.5 M, pH 8.5) - 250ml
Supplier:  Boston Bioproducts Inc BB66250ML
Encompass
The Borate Buffer is an isotonic buffer essential for maintaining stable pH levels in biochemical processes. Typically operating within a pH range of 8.0 to 9.5, this buffer supports several applications including enzymatic assays, protein purification, as a DNA electrophoresis buffer, in coating procedures, and for coupling proteins. The Borate Buffer is also used as plant tissue culture media, and has a strong bactericidal effect.
Catalog No. NC2151132
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.