Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Borate Buffer, pH 9.5, for Ammonia and Organic Nitrogen Analysis, Reagents

Catalog No. 01335830
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01-335-830 4 L Poly Bottle
01-335-828 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-335-830 Supplier Reagents Holdings Llc Supplier No. CB0095004A
Add to Cart
Add to Cart

Borate Buffer, pH 9.5, for Ammonia and Organic Nitrogen Analysis, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Buffer
CAS 7732-18-5
Color Colorless
pH 9.5
Physical Form Liquid
Quantity 4 L
Packaging Poly Bottle
Sterility Non-sterile
Solubility Information Completely miscible
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.