Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Borax Standard Buffer, Fisher Chemical™
SDP

Catalog No. sb9200500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SB9200500 500 mL
SB92001 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SB9200500
Only null left
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Borax Standard Buffer
CAS 7732.18-5, 1303-96-4
For Use With (Application) For AOAC Method 946.01
Physical Form Solution
Quantity 500 mL
Packaging Natural Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.