Learn More
ATCC Bordetella pertussis Strain D946 (ATCC BAA-3221)
Supplier: ATCC BAA3221
Bordetella pertussis strain D946 (ATCC BAA-3221) is a preserved reference culture intended for research and reference use. The strain is supplied as a preserved culture with defined storage and shipping conditions, and it requires Biosafety Level 2 handling.
- Preserved reference bacterial culture for research and reference use.
- Organism: Bordetella pertussis.
- Strain: D946.
- Storage temperature: liquid nitrogen.
- Shipping temperature: -80°C.
- Product format: preserved culture, single solution.
- Biosafety level: 2.
- Country of origin: United States.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.