Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boric Acid, 2% (w/v), Ricca Chemical
SDP

Catalog No. R10650004F Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R10650004F 4 L Cubitainer
10658 250 mL Natural Poly Bottle
106516 500 mL Natural Poly Bottle
1065-32 1 L Natural Poly Bottle
1065-1 4 L Natural Poly Bottle
R106500010F 10 L Cubitainer
10655 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. R10650004F Supplier Ricca Chemical Company Supplier No. 10651CT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7732-18-5 , 10043-35-3
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
Synonym orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
PubChem CID 7628
ChEBI CHEBI:33118
IUPAC Name boric acid
SMILES OB(O)O

Specifications

Color Colorless
Concentration 2% (w/v)
CAS Min % 96.0, 1.92
CAS Max % 100.0, 2.0
Packaging Cubitainer
Solvent or Matrix Aqueous Solution
Quantity 4 L
Physical Form Liquid
Percent Purity 2% W/V
Grade Laboratory
Chemical Name or Material Boric Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.