Learn More
Boric Acid, 2% (w/v) with Indicator (Methylene Blue-Methyl Red) for Nitrogen (Ammonia), Nitrogen (Organic), Certified, LabChem™
Shop All LabChem ProductsSpecifications
Specifications
| CAS Max % | 2, 96, 1, 0.9, 0.052 |
| Identification | Passes Test |
| Packaging | Poly Bottle |
| Suitability | Passes Test |
| Linear Formula | H3BO3 |
| Decomposition Information | Boron; Carbon monoxide; Carbon dioxide |
| Color | Purple |
| Solubility Information | Soluble in water |
| Quantity | 1 L |
| Physical Form | Liquid |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement May damage fertility or the unborn child. |
| Precautionary Statement | GHS P Statement Obtain special instructions before use. Do not handle until all safety precautions have been read and understood. Wear protective gloves, eye protection. If exposed or concerned: Get medical advice/attention. Store locked up. Dispose of contents/container to comply with local, state and federal regulations. |
| Recommended Storage | Room Temperature |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.