Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boric Acid, 4% (w/v), Ricca Chemical
SDP

Catalog No. R106800020F Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R106800020F 20 L
106816 500 mL
1068-32 1 L
1068-1 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. R106800020F Supplier Ricca Chemical Company Supplier No. R106800020F
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7732-18-5 , 10043-35-3
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
PubChem CID 7628
ChEBI CHEBI:33118
IUPAC Name boric acid
SMILES OB(O)O

Specifications

Appearance of Solution Colorless Liquid
CAS Min % 94.0, 3.77
CAS Max % 98.0, 3.93
Packaging Cubitainer
Quantity 20 L
Synonym orthoboric acid
Percent Purity 4% W/V
Grade Laboratory
Chemical Name or Material Boric Acid Solution, 4%(w/v) Aqueous Solution
ShelfLife 24 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.