Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boric Acid, ≥ 99.0%, Suprapur™, MilliporeSigma™

Catalog No. M1007650500 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1007650500 500 g
M1007650050 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1007650500 Supplier MilliporeSigma Supplier No. 1007650500
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10043-35-3
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
Synonym Orthoboric acid, Trihydroxidoboron
IUPAC Name boric acid
SMILES OB(O)O

Specifications

pH 3.8 to 4.8 (33 g/L, H2O, 20°C)
Density 1.489 g/cm3 (at 23°C)
Packaging Plastic Bottle
Vapor Pressure < 1 Pa (25°C)
Assay Percent Range ≥ 99.0%
Quantity 500 g
Solubility Information 49.2 g/L
Percent Purity ≥99.9999%
Recommended Storage Store at 5 to 30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.