Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boric Acid, 3% (w/v) with Mixed Indicator, Ricca Chemical
SDP

Catalog No. 10675 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10675 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10675 Supplier Ricca Chemical Company Supplier No. 10675
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5 , 10043-35-3 , 62625-32-5 , 845-10-3
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
Synonym orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
PubChem CID 7628
ChEBI CHEBI:33118
IUPAC Name boric acid
SMILES OB(O)O

Specifications

Appearance Clear reddish-orange colored liquid
Odor Odorless
Packaging Cubitainer
Assay Percent Range 3% (w/v)
Quantity 20 L
Physical Form Solution
Solubility Information Solubility in Water: Infinite
Percent Purity 3% W/V
Grade Laboratory
Chemical Name or Material Boric Acid
ShelfLife 18 months
Recommended Storage General
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.