Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boric Acid, Certified, 2.0% (w/v) ±0.1%, LabChem™

Catalog No. LC117202
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC117202 1 L
LC117204 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC117202 Supplier LabChem, Inc. Supplier No. LC117202
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10043-35-3 , 7732-18-5
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
Synonym orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
PubChem CID 7628
ChEBI CHEBI:33118
IUPAC Name boric acid
SMILES OB(O)O

Specifications

Color Colorless
CAS Max % 98, 2
Identification Passes Test
Packaging Poly Bottle
Traceability to NIST Traceable to NIST
Decomposition Information Boron
Quantity 1 L
Physical Form Liquid
Linear Formula H3BO3
Solubility Information Soluble in water
Formula Weight 61.83
Percent Purity 2.0% ±0.1% (w/v)
Grade Certified
Chemical Name or Material Boric Acid
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
May damage fertility or the unborn child.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves, eye protection.
If exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.