Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boric Acid, Granular, ACS, 99.5%, Spectrum™ Chemical
SDP

Catalog No. 18609171 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-609-171 500 g Poly Bottle
18-609-169 125 g Poly Bottle
18-609-167 2.5 kg Poly Bottle
18-609-170 12 kg Poly Pail
18-609-168 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-609-171 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. B1131500GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10043-35-3
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
IUPAC Name boric acid
SMILES OB(O)O

Specifications

CAS Max % 1
Heavy Metals 1e-05
Packaging Poly Bottle
Quantity 500 g
Percent Purity 99.5%
Grade ACS

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.