Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boric Acid, Granular, BiotechGrade, 99.5-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18601045 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
50 kg
Packaging:
Fiber Drum
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-045 500 g Poly Bottle
18-601-046 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-601-045 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. B1129500GM
Add to Cart
Add to Cart

B1129, 10043-35-3, H3BO3

Spectrum™ Chemical Boric Acid, Granular, BiotechGrade , is a weak acid of boron often used as an antiseptic, insecticide, flame retardant, neutron absorber, or precursor to other chemical compounds. It exists in the form of colorless crystals or a white powder that dissolves in water. Boric acid is poisonous if taken internally or inhaled in large quantities.

Chemical Identifiers

CAS 10043-35-3
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
IUPAC Name boric acid
SMILES OB(O)O

Specifications

pH 3.8-4.8
CAS Max % 100%
Heavy Metals 0.002%
Packaging Poly Bottle
Quantity 500 g
Percent Purity 99.5 to 100.5%
Grade Biotech Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.