Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boric Acid, Lab Grade Crystals, Innovating Science™
SCI_ED

Catalog No. S000542 Shop All Aldon Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000542 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger: May damage fertility or the unborn child.
Catalog No. S000542 Supplier Aldon Corporation Supplier No. IS11055
Add to Cart
Add to Cart
Danger: May damage fertility or the unborn child.

Chemical Identifiers

CAS 10043-35-3
Molecular Weight (g/mol) 61.83 g/mol

Specifications

Quantity 100 g
Percent Purity 100%
Grade Laboratory
Chemical Name or Material Boric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.