Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boric Acid, MP Biomedicals™

Catalog No. MP021914113 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021914113 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP021914113 Supplier MP Biomedicals, Inc Supplier No. 02191411.3
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used in buffers and for calibration of gel columns, as well as an astringent, antiseptic, antifungal and antibacterial agent. Boric Acid, MP Biomedicals has a single boron and its salt forms complex equilibria in solution, with the ion B4O7-2 being the pr

Application include:

  • As an astringent, antiseptic, antibacterial and antifungal agent in research labs
  • In buffers and for calibration of polyacrylamide gel columns for the separation of oligonucleotides by capillary electrophoresis
  • Weatherproofing wood and fireproofing fabrics
  • Manufacturing of cements, crockery, porcelain, enamels, glass, borates, leather, carpets, hats, soaps, artificial gems
  • As a preservative and in nickeling baths
  • Cosmetics, printing, dyeing, painting and photography
  • For impregnating wicks, in electric condensers and for hardening steel
  • As an insecticide for cockroaches and black carpet beetles
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10043-35-3
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
Synonym orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
PubChem CID 7628
ChEBI CHEBI:33118
IUPAC Name boric acid
SMILES OB(O)O

Specifications

Quantity 25 g
Physical Form White Powder
Solubility Information Soluble in water; insoluble in alcohol; freely soluble in boiling water.
Formula Weight 61.84g/mol
Percent Purity ≥99.5%
Grade ACS Reagent
Recommended Storage Room Temeprature; Dessicate
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.