Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boric Acid, MP Biomedicals™

Catalog No. MP021914111 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021914111 100 g
MP021914115 500 g
MP219141101 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP021914111 Supplier MP Biomedicals, Inc Supplier No. 02191411.1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used as an astringent, antiseptic, antibacterial and antifungal agent. Boric Acid, MP Biomedicals is a preservative with a wide range of applications. It is used in buffers and for calibration of polyacrylamide gel columns for the separation of oligonucle

Boric acid has a single boron; its salt forms complex equilibria in solution, with the ion B4O7-2 being the predominant species. Manufacturing and industrial uses include: for weatherproofing wood and fireproofing fabrics; as a preservative; manufacturing of cements, crockery, porcelain, enamels, glass, borates, leather, carpets, hats, soaps, artificial gems; in nickeling baths; cosmetics; printing and dyeing, painting; photography; for impregnating wicks; electric condensers; hardening steel. It is also used as an insecticide for cockroaches and black carpet beetles.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10043-35-3
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
Synonym orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
PubChem CID 7628
ChEBI CHEBI:33118
IUPAC Name boric acid
SMILES OB(O)O

Specifications

Quantity 100 g
Physical Form White Powder
Solubility Information Soluble in water; insoluble in alcohol; freely soluble in boiling water.
Formula Weight 61.84
Percent Purity ≥99.5%
Grade ACS Reagent
Recommended Storage Room Temperature (15-30°C), dessicate.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.