Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boric Anhydride, Purified, 97%, Spectrum™ Chemical
SDP

Catalog No. 18609188 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-609-188 12 kg Poly Bottle
18-609-187 125 g Poly Bottle
18-609-189 500 g Poly Bottle
18-609-186 2.5 kg Poly Bottle
18-609-190 50 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-609-188 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. B113512KG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1303-86-2

Specifications

CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 12 kg
Percent Purity 97%
Grade Purified
Packaging Poly Bottle
Chemical Name or Material Boric Anhydride

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.