Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boron ICP Standard, 100 ppm B in H2O, Ricca Chemical
SDP

Catalog No. RPB100W0100 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RPB100W0100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RPB100W0100 Supplier Ricca Chemical Company Supplier No. RPB100W0100N
Add to Cart
Add to Cart

Single Element AA Standard

Chemical Identifiers

CAS 10043-35-3 , 7732-18-5
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
PubChem CID 7628
ChEBI CHEBI:33118
IUPAC Name boric acid
SMILES OB(O)O

Specifications

Packaging Natural LDPE Bottle
CAS Min % 99.9, 0.1
CAS Max % 99.9, 0.1
Density 1
Physical Form Liquid
Color Colorless
Melting Point 0°C
Boiling Point 100°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type Standard
Quantity 100 mL
Grade Laboratory
Chemical Name or Material Boron ICP Standard, 100 ppm B in H2O
Show More Show Less
ShelfLife 12 months
Recommended Storage Ambient
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.