Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boron powder, amorphous, -325 mesh, 90%, Mg nominal 5%, Thermo Scientific Chemicals

Catalog No. AA1133814
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1133814 25 g
AA1133822 100 g
AA1133836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1133814 Supplier Thermo Scientific Chemicals Supplier No. 1133814

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-42-8
Molecular Formula B
Molecular Weight (g/mol) 10.81
MDL Number MFCD00134034 MFCD00151272
InChI Key ZOXJGFHDIHLPTG-UHFFFAOYSA-N
Synonym boron, metallic, unii-n9e3x5056q, boron, amorphous, powder, crystalline, boro, boron-elemental, boron, elemental, and compounds, boracium, boranylidyne
PubChem CID 5462311
ChEBI CHEBI:27560
IUPAC Name boron
SMILES [B]

Specifications

Quantity 25 g
Mesh Size −325 Mesh
Odor Odorless
Form Powder
UN Number UN3178
Percent Purity 90%
Concentration or Composition (by Analyte or Components) Mg nomin.al 5%
Chemical Name or Material Boron powder, Amorphous
Hazard Category Warning
Hazard Statement GHS H Statement
H228-H302-H332-H335
Flammable solid.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P210-P261-P280-P304+P340-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.
EINECSNumber 231-151-2
RTECSNumber ED7350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.