Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boron Trifluoride - Ethyl Ether Complex 98.0+%, TCI America™
SDP

Catalog No. B0527100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0527100ML 100 mL
B052725ML 25 mL
B0527500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. B0527100ML Supplier TCI America Supplier No. B0527100ML
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 109-63-7
Molecular Formula C4H10BF3O
Molecular Weight (g/mol) 141.93
MDL Number MFCD00013194
InChI Key KZMGYPLQYOPHEL-UHFFFAOYSA-N
PubChem CID 8000
IUPAC Name ethoxyethane; trifluoroborane
SMILES FB(F)F.CCOCC

Specifications

Melting Point -60°C
Color Yellow
Boiling Point 124°C
Quantity 100 mL
UN Number 2604
Formula Weight 141.93
Percent Purity ≥98.0% (W)
Physical Form Liquid
Chemical Name or Material Boron Trifluoride - Ethyl Ether Complex
EINECSNumber (2)-2107
RTECSNumber ED2275000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.