Learn More
Bellco Glass, Inc. Borosil Pipettes, Measuring (Mohr), Class A, 2.0mL x 0.02mL, Batch Cert
SDP
Supplier:  Bellco Glass, Inc. 452801073
Encompass_Preferred
DescriptionBorosil Graduated Mohr Pipette Class A Batch Certificate, EN ISO 835, 3.3 Borosilicate glass, Amber markings, graduated from zero to last graduated mark, To deliver, CS/10, 2mL (2*0.02)SpecificationsMaterial: 3.3 Borosilicate Glass Color: ClearCapacity: 2mLOverall Dimension: 18.70" L x 18.70" W x 6.89" HGrad. Interval: .02Tolerance: .01Class/Quality Grade: Type 1, Class A
Catalog No. 50-254-9689