Learn More
Bellco Glass, Inc. Borosil Tubes, Culture, Flat Bottom, Amber, PTFE-Lined PP Screw Caps, 15mL
Supplier: Bellco Glass, Inc. 452800900
Borosil Amber Culture Tubes Media Flat Bottom PP cap with Liner 15 mL Borosilicate CS/100SpecificationsMaterial: 3.3 Borosilicate Glass Tube size: 15 mLColor: AmberCap Material: Polypropylene Overall Dimension: 9911007Class/Quality Grade: Type 1, Class A