Learn More
Eisco Labs Borosilicate Glass Erlenmeyer Flask - ASTM, Dual Graduated Scale
Catalog No. 12141677 Shop All Eisco Products
Change view
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
1000 mL
2000 mL
4000 mL
Mouth Style:
Narrow
Wide
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Mouth Style
12-141-677 1000 mL Narrow
12-141-638 125 mL Narrow
12-141-675 125 mL Wide
12-141-659 250 mL Narrow
12-141-623 250 mL Wide
12-141-676 500 mL Narrow
12-141-624 500 mL Wide
12-141-625 1000 mL Wide
12-141-642 4000 mL Narrow
12-141-657 2000 mL Wide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 12-141-677 Supplier Eisco Labs Supplier No. CH2162
Only null left
Add to Cart
Add to Cart
Durable conical design ideal for mixing, heating, and storing liquids in laboratory settings.
  • Industrial Quality :
    • Conical Flask made from high quality, heavy duty borosilicate 3.3 glass conforming to USP Type-1 and ASTM E438 Type-1, Class A requirements.
    • Excellent chemical resistance and strength against breakage.
  • Superior Durability :
    • Features uniform wall thickness for ideal heating.
    • It is microwavable and autoclavable, and can withstand high temperatures and thermal shocks.
    • The flask has a reinforced rim to protect against high impacts.
  • Dual Graduations :
    • Easy-to-read white enamel dual graduated scale for added accuracy when filling or dispensing
  • Meets ASTM E1404 Standards :
    • Conforms with ASTM Specification E1404, Type-1, Class-1 requirements for glass conical flasks.
Specifications
Capacity (Metric) 1000 mL
Material Borosilicate Glass (3.3)
Product Type Erlenmeyer Flask
Autoclavable Autoclavable
Disposable Reusable
Graduated Yes
Mouth Style Narrow
Sterility Non-sterile
Bottom Shape Flat
Surface Treatment Non-treated