Learn More
United Scientific™ Borosilicate Glass Low Form Heavy Duty Beakers Set of 3
SDPSCI_ED
Catalog No. S111031 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Autoclavable:
Autoclavable
Graduated:
Yes
Material:
Borosilicate Glass (3.3)
Style:
Heavy Wall
Catalog No. Autoclavable Graduated Material Style
S111031 Autoclavable Yes Borosilicate Glass (3.3) Heavy Wall
1 options
Catalog No. S111031 Supplier United Scientific™ Supplier No. BGSET3HD
Add to Cart
Add to Cart
These heavy duty beakers are ideal for tough handling conditions including mechanized washing. Beakers feature a double graduated metric scale and a marking area.
  • Heavy-duty beaker set of 3 includes 250 mL, 400 mL, and 600 mL.
  • Made from low-expansion borosilicate glass with extra-thick walls, this beaker is ideal for tough handling conditions, including mechanized washing.
  • Also includes white printed graduations and a marking area.
  • Tolerance is approximately ±5%.
Specifications
Capacity (Metric) 250 mL, 400 mL, 600 mL
Material Borosilicate Glass (3.3)
For Use With (Application) General Lab Use
Autoclavable Autoclavable
Disposable Reusable
Graduated Yes
Scale White Double Scale
Style Heavy Wall