Learn More
Eisco™ Borosilicate Prism
SCI_ED
Catalog No. S14037 Shop All Eisco Products
Change view
Click to view available options
Refractive index: 1.51
Catalog No.
S14037 Refractive index: 1.51
1 options
Catalog No. S14037 Supplier Eisco™ Supplier No. PH0583A
Add to Cart
Add to Cart
Designed for use with spectrometer.
  • Borosilicate crown glass
  • Refractive index of 1.51
Specifications
Product Type Borosilicate Prism
Description Refractive index: 1.51
Material Borosilicate Crown Glass