Learn More
Creative Biomart Recombinant Borrelia Afzelii OspA Antigen, His-tagged
Supplier:  Creative Biomart BAO003
Encompass_Preferred
Recombinant Borrelia Afzelii Outer Surface Protein A produced in E. coli is a non-glycosylated, polypeptide chain having a calculated molecular mass of 29,074 Dalton.Borrelia Afzelii OspA is expressed
Catalog No. 50-208-4381
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.