Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Borrelia burgdorferi Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA173004 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA173004 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA173004 Supplier Invitrogen™ Supplier No. PA173004
  • $626.00
    $479.65 / Each of 1

    Save $146.35

    23% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

The PA1-73004 antibody reacts with Borrelia burgdorferi. This antibody cross-reacts with Treponema pallidum, B. hermsii & B. parkerii. PA1-73004 has been successfully used in Western blot, Immunohistochemistry (paraffin), and immunofluorescence applications. By Western blot, this antibody detects bands at 83, 41, 34 & 31 kDa, plus additional low MW bands. For immunofluorescence, specimens should be acetone-fixed for best results. The PA1-73004 antibody was raised against B. burgdorferi whole cell preparation. Short term, less than 6 months, storage is liquid at 4°C. For longer term storage, aliquot and freeze.

Borrelia burgdorferi is a spirochete and the cause of Lyme disease, a tick transmitted illness of humans and animals. B. burgdorferi may persist in humans and animals for months or years following initial infection, despite a robust humoral immune response. B. burgdorferi resembles other spirochetes in that it is a highly specialized, motile, two-membrane, spiral shaped bacteria which lives primarily as an extracellular pathogen. B. burgdorferi has an unusual genome compared with other eubacteria which includes a linear chromosome approximately one megabase in size and numerous linear and circular plasmids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Borrelia burgdorferi
Applications Western Blot
Classification Polyclonal
Concentration 5.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias B. burgdorferi; Borrelia; Lyme disease
Host Species Rabbit
Immunogen B. burgdorferi whole cell preparation.
Purification Method Protein A
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.