Learn More
Creative Biomart Recombinant Borrelia NapA Antigen, His-tagged
Supplier:  Creative Biomart BNA015
Encompass_Preferred
Recombinant Borrelia Burgdorferi NapA produced in E. coli is a non-glycosylated, full-length polypeptide chain having a calculated molecular mass of 22,094 Dalton.Borrelia NapA is expressed with a 6
Catalog No. 50-208-4393
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.