Learn More
Creative Biomart Recombinant Borrelia p41 Antigen, His-tagged
Supplier:  Creative Biomart BP1019
Encompass_Preferred
The E. coli derived recombinant protein contains the p41 immunodominant regions, 158-296 amino acids. Borrelia p41 is fused to 6 His tag at N-terminal and purified by proprietary chromatographic te
Catalog No. 50-208-4397
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.