Learn More
Creative Biomart Recombinant Borrelia VisE1 Antigen, His-tagged
Supplier:  Creative Biomart BVE026
Encompass_Preferred
Recombinant Borrelia VisE1 (variable major protein like sequence E1) produced in E. coli is a non-glycosylated, polypeptide chain having a calculated molecular mass of 43 kDa.Borrelia VisE1 is express
Catalog No. 50-208-4404
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.