Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Bortezomib
GREENER_CHOICE

Catalog No. p-7235823
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
72-822-5 25 mg
72-825 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 72-822-5 Supplier Tocris Bioscience™ Supplier No. 7282/25
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

High affinity proteasome inhibitor

Bortezomib is a high affinity proteasome inhibitor (Ki = 0.6 nM). Inhibits growth of multiple myeloma (MM) cell lines (IC50 values in <50 nM range); induces apoptosis in MM cell lines and MM patient derived cells, including p53 mutant cell lines. Decreases adherence of MM cells lines to bone marrow stromal cells (BMSCs). Acts synergistically with dexamethasone (Cat. No. 1126). Also cytotoxic in MCF-7 breast cancer cell line (IC50 = 50 nM).

Specifications

Chemical Name or Material B-[(1 R)-3-Methyl-1-[[(2 S)-1-oxo-3-phenyl-2-[(2-pyrazinylcarbonyl)amino]propyl]amino]butyl]boronic acid
CAS 179324-69-7
Quantity 25 mg
Target Proteasome Inhibitors
Molecular Formula C19H25BN4O4
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.