Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Bortezomib, Proteasome Inhibitor, Solid, >98% Purity, 25 mg

Catalog No. 502583326
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-3326 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-3326 Supplier Abcam Supplier No. AB14212325MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent, selective, and reversible proteasome inhibitor with greater than 98% purity.

  • Potency: Potent, selective, reversible proteasome inhibitor (Ki = 0.6 nM).
  • Biological Activity: Inhibits proliferation of tumor cell lines (IC50 = 7 nM), TNF synthesis, and FGF-induced angiogenesis.
  • Purity: >98%.
  • Molecular Weight: 384.2 Da.
  • Form: Solid.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Substrate Properties Chemical Substrate
Target Molecule Proteasome
Content And Storage Store at -20°C.
Format Vial
Form Solid
Shipping Condition Ambient
Quantity 25 mg
For Use With (Application) Proteasome inhibition, Cancer-proliferation inhibition, Angiogenesis inhibition
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.