Learn More
Apexbio Technology LLC SU11274 658084-23-2 100mg
SDP
Supplier:  Apexbio Technology LLC A2678100
Encompass_Preferred
SU11274 (CAS 658084-23-2) is a selective inhibitor targeting Met kinase exhibiting an IC50 of approximately 10 nM The compound specifically suppresses the autophosphorylation of Met at residues Tyr1234/1235 disrupting receptor activity with minimal effects on other receptor tyrosine kinases such as EGFR and PDGFR or serine/threonine kinases including CDK2 In NIH3T3 cells expressing drug-sensitive or resistant MET mutants and in H1993 human lung cancer cells treatment with SU11274 induces a dose-dependent inhibition of Met phosphorylation and impairs receptor ubiquitination leading to receptor accumulation SU11274 serves as a useful tool for investigating Met signaling pathways in oncology research
Catalog No. 50-000-63253
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.