Learn More
Selleck Chemical LLC Bortezomib (PS-341) 5mg 179324-69-7 LDP-341, MLM341
SDP
Supplier:  Selleck Chemical LLC S10135MG
Encompass_Preferred
Bortezomib (PS-341, LDP-341, MLM341) is a potent 20S proteasome inhibitor with Ki of 0.6 nM. It exhibits favorable selectivity towards tumor cells over normal cells. Bortezomib (PS-341) inhibits NF-?B and induces ERK phosphorylation to suppress cathepsin B and inhibit the catalytic process of autophagy in ovarian cancer and other solid tumors. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-741-9
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.