Learn More
Medchemexpress LLC HY-A0013 500mg Medchemexpress, Bosentan CAS:147536-97-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYA0013500MG
Encompass_Preferred
Medchemexpress, HY-A0013 500mg Bosentan CAS:147536-97-8 Bosentan is a competitive and dual antagonist of endothelin-1 (ET) for the ETA and ETB receptors with Ki of 4.7 nM and 95 nM in human SMC, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9375
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.