Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Boss Head
SCI_ED

Catalog No. S24532 Shop All Eisco Products
Change view
Click to view available options
Color:
Gray
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S24532 Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S24532 Supplier Eisco™ Supplier No. CH0663AP
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with laboratory stands.

  • Made from die-cast alloy
  • Accepts rods up to 16mm dia. at right angles
  • Metal thumbscrews with blue plastic heads

Specifications

Type Bosshead Clamp
Material Die Cast Alloy
For Use With (Equipment) Accepts rods up to 16mm dia.
Size 11.7 x 3 x 3cm
Color Gray
Jaw Type Fixed
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.