Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Protective Industrial Products Boss™ Three-Piece Rainsuit

Catalog No. 19036613 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
7X-Large
8X-Large
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-036-613 Small
19-036-612 Medium
19-036-611 Large
19-036-614 X-Large
19-036-615 2X-Large
19-036-616 3X-Large
19-036-617 4X-Large
19-036-618 5X-Large
19-036-619 6X-Large
19-036-609 7X-Large
19-036-610 8X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Catalog No. 19-036-613 Supplier Protective Industrial Products Supplier No. 4035/S
Add to Cart
Add to Cart
This item is not returnable. View return policy

Boss™ Three-Piece Rainsuit. Ideal for construction, municipalities, shipyards, agriculture chemical spraying and industrial plant maintenance.

  • Made with 0.35 mm thick PVC and polyester fabric
  • Detachable hood with drawstring|Storm flap front with snap closure
  • 2 flapped front pockets with snap closures to secure valuables
  • Cape vented back
  • Corduroy collar for comfort
  • Adjustable suspenders for with triangular buckles for a perfect fit
  • Front fly opening with snap

Specifications

Color Yellow
For Use With (Application) Construction, Municipalities, Shipyards, Agriculture Chemical Spraying and Industrial Plant Maintenance.
Hood Type Detachable with Drawstring
Material PVC
Size Small

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.