Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synthware™ Bosshead
SDP

Catalog No. 31502241 Shop All Kemtech Products
Change view
Click to view available options
Size (Metric) Grip:
15 mm
16 mm
19 mm
Style:
Flat Knobs
Knobless
Round Knobs
Wing Nuts
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size (Metric) Grip Style
31-502-241 16 mm Flat Knobs
31-502-240 15 mm Wing Nuts
31-501-957 15 mm Round Knobs
31-502-242 19 mm Knobless
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 31-502-241 Supplier Synthware™ Supplier No. W200002B
Add to Cart
Add to Cart

Bosshead for attaching clamps to hood rack system

Designed for use with rods ranging from 15 to 19mm in diameter.

Specifications

Style Flat Knobs
Type Bosshead Clamp
Material Chrome-plated Metal
Includes Bosshead only
For Use With (Application) Organic Synthesis
Description Flat metal knobs
Size Fits 16 mm Rod
Size (Metric) Grip 16 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.