Learn More
R&D Systems™ 10X SUMO Conjugation Reaction Buffer
Description
Specifications
Specifications
| Product Line | R&D Systems |
| Quantity | 5 x 1 mL |
| Storage Requirements | Store the unopened product at -20° to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |