Learn More
R&D Systems™ 20X 20S Proteasome Reaction Buffer
SDP
Catalog No. B80R Shop All Novus Products
Change view
Click to view available options
Quantity:
2 x 1 mL
Catalog No. Quantity
B80R 2 x 1 mL
1 options
This item is not returnable. View return policy
Catalog No. B80R Supplier R&D Systems™ Supplier No. B80
Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
Contains the optimal formulation for the quantitative analysis of constitutive 20S and immuno 20S proteasome activity
20S Reaction Buffer contains the optimal formulation for the quantitative analysis of constitutive 20S and immuno 20S proteasome activity. This buffer can be used as supplement to the 20S Proteasome Assay Kit (K-900) or with any individual proteasome enzymes (E-350, E-360, E-370, E-375) in conjunction with the fluorogenic peptide Suc-Leu-Leu-Val-Tyr-AMC (S-280) and 20S proteasome activation solution (B-30) to measure activity.
Specifications
Product Line R&D Systems
Quantity 2 x 1 mL
Storage Requirements Store the unopened product at -20° to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.